• USA Home
  • Search Results

  • chlorhexidinedihydrochloride57837369742511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorhexidine dihydrochloride

2 Products
Synonym:   1,6-Bis(N5-[p-chlorophenyl]-N1-biguanido)hexane; 1,1′-Hexamethylenebis(5-[p-chlorophenyl]biguanide), Lisium
Image of Chlorhexidine dihydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C22H30Cl2N10 · 2HCl
Molecular Weight:
 578.37
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More