• USA Home
  • Search Results

  • chloro13bis246trimethylphenylimidazol2ylidenegoldi5368585244581911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloro[1,3-bis(2,4,6-trimethylphenyl)imidazol-2-ylidene]gold(I)

1 Products
Synonym:   1,3-Bis(2,4,6-trimethylphenyl)imidazol-2-ylidene gold(I) chloride
Image of Chloro[1,3-<WBR>bis(2,4,6-<WBR>trimethylphenyl)<WBR>imidazol-<WBR>2-<WBR>ylidene]<WBR>gold(I)
CAS Number:
Empirical Formula (Hill Notation):
 C21H24AuClN2
Molecular Weight:
 536.85
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More