• USA Home
  • Search Results

  • chloro15cyclooctadienerhodiumidimer493081209247611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloro(1,5-cyclooctadiene)rhodium(I) dimer

2 Products
Synonym:   1,5-Cyclooctadienerhodium(I) chloride dimer, Bis(1,5-cyclooctadiene)dirhodium(I) dichloride, Di-μ-chlorobis[(1,2,5,6-η)-1,5-cyclooctadiene]dirhodium, Rhodium(I) chloride 1,5-Cyclooctadiene complex dimer, [Rh(COD)Cl]2
Image of Chloro(1,5-<WBR>cyclooctadiene)<WBR>rhodium(I) dimer
CAS Number:
Empirical Formula (Hill Notation):
 C16H24Cl2Rh2
Molecular Weight:
 493.08
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Reaction Suitability
Show More