• USA Home
  • Search Results

  • chloroaceticacid94507911811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloroacetic acid

6 Products
Synonym:   Monochloroacetic acid
Image of Chloroacetic acid
CAS Number:
Linear Formula:
 ClCH2COOH
Molecular Weight:
 94.50
 Beilstein/REAXYS Number:
 605438
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Physical Form
  Purity Grade
  Potency
  Solubility
  Melting Point
  Special Grade
  Application(s)
Show More