• USA Home
  • Search Results

  • chloroacetone92527895511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloroacetone

3 Products
Synonym:   1-Chloropropan-2-one, Chloro-2-propanone, MCA
Image of Chloroacetone
CAS Number:
Linear Formula:
 ClCH2COCH3
Molecular Weight:
 92.52
 Beilstein/REAXYS Number:
 605369
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Additive
  Assay
Show More