• USA Home
  • Search Results

  • chlorobenzene1125610890711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorobenzene

12 Products
Image of Chlorobenzene
CAS Number:
Empirical Formula (Hill Notation):
 C6H5Cl
Molecular Weight:
 112.56
 Beilstein/REAXYS Number:
 605632
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Restriction
  Refractive Index
  Solubility
  Potency
  Package quantity
  Explosion limit
  Autoignition temp
  Vapor pressure
  Transition temperature
  Application(s)
  Product Line
  Melting Point
  Density
  UV-Absorption Values
  pH Value
  Physical Form
  Special Grade
Show More