• USA Home
  • Search Results

  • chlorobenzenesolution1125610890711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorobenzene solution

1 Products
CAS Number:
Empirical Formula (Hill Notation):
 C6H5Cl
Molecular Weight:
 112.56
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More