• USA Home
  • Search Results

  • chlorodimethylphenylsilane1707176833211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloro(dimethyl)phenylsilane

1 Products
Synonym:   DMPSCl, Dimethylphenylchlorosilane, Dimethylphenylsilylchloride, Phenyldimethylchlorosilane
Image of Chloro(dimethyl)<WBR>phenylsilane
CAS Number:
Linear Formula:
 C6H5Si(CH3)2Cl
Molecular Weight:
 170.71
 Beilstein/REAXYS Number:
 606292
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More