• USA Home
  • Search Results

  • chloroethanesolution64517500311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloroethane solution

3 Products
Synonym:   Ethyl chloride
Image of Chloroethane solution
CAS Number:
Empirical Formula (Hill Notation):
 C2H5Cl
Molecular Weight:
 64.51
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Titertest
  Density
  Anal. Standard Form
  Package quantity
  Vapor pressure
  Special Grade
  Concentration
Show More