• USA Home
  • Search Results

  • chloroform119386766311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloroform

36 Products
Synonym:   Methylidyne trichloride, Trichloromethane
Image of Chloroform
CAS Number:
Empirical Formula (Hill Notation):
 CHCl3
Molecular Weight:
 119.38
 Beilstein/REAXYS Number:
 1731042
 EC Number:
 MDL number:
 NACRES:
 NA.04
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Purification method
  Refractive Index
  Absorption
  Quality clarifier
  Purity Grade
  Restriction
  Transmittance
  Product Line
  Ice Shipping
  Agency/Method
  UV-Absorption Condition
  Solubility
  Package quantity
  Suitability
  Potency
  Density
  Color
  Additive
  Special Grade
  UV-Absorption Values
  Application(s)
  Physical Form
Show More

Product #
Description
C2432 Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%,
288306 Chloroform, anhydrous, ≥99%, contains 0.5-1.0% ethanol as stabilizer,
372978 Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%,
650498 Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer,
366927 Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer,
32211 Chloroform, puriss. p.a., reag. ISO, reag. Ph. Eur., 99.0-99.4% (GC),
34854 Chloroform, suitable for HPLC, ≥99.8%, amylene stabilized,
319988 Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%,
1.02445 Chloroform, for analysis EMSURE® ACS,ISO,Reag. Ph Eur,
472476 Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%,
132950 Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer,
366919 Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer,
02487 Chloroform, analytical standard,
650471 Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer,
496189 Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer,
1.02444 Chloroform, for liquid chromatography LiChrosolv®,
PHR1552 Chloroform, Pharmaceutical Secondary Standard; Certified Reference Material,
CX1059 Chloroform, OmniSolv®,
C7559 Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer,
1.02432 Chloroform, for gas chromatography ECD and FID SupraSolv®,
1.02447 Chloroform, for spectroscopy Uvasol®,
8.22265 Chloroform, EMPLURA®,
24216 Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC),
1.07024 Chloroform, for analysis EMPARTA® ACS,
1.02431 Chloroform, EMPROVE® ESSENTIAL,
CX1055 Chloroform, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS,
48520-U Chloroform, analytical standard,
1.02442 Chloroform, for analysis (for determinations with dithizone),
480150 Chloroform, ACS reagent, ≥99.8%, contains amylenes as stabilizer,
CX1050 Chloroform, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs,
439142 Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer,
437581 Chloroform, ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer,
CX1054 Chloroform, OmniSolv®,
CX1058 Chloroform, HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs,
CX1060 Chloroform,
270636 Chloroform, suitable for HPLC, ≥99.8%,