• USA Home
  • Search Results

  • chloroformd1203886549611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloroform-d

11 Products
Synonym:   Deuterochloroform
Image of Chloroform-d
CAS Number:
Linear Formula:
 CDCl3
Molecular Weight:
 120.38
 Beilstein/REAXYS Number:
 1697633
 EC Number:
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Isotopic Purity
  Special Grade
  Package quantity
  Application(s)
  Additive
  Physical Form
  Assay
Show More