• USA Home
  • Search Results

  • chlorogenicacid3543132797911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorogenic acid

6 Products
Synonym:   1,4,5-Trihydroxycyclohexanecarboxylic acid 3-(3,4-dihydroxycinnamate), 3-(3,4-Dihydroxycinnamoyl)quinic acid, CGA
Image of Chlorogenic acid
CAS Number:
Empirical Formula (Hill Notation):
 C16H18O9
Molecular Weight:
 354.31
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Assay
Show More