• USA Home
  • Search Results

  • chloropentafluorobenzene2025134407011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloropentafluorobenzene

1 Products
Image of Chloropentafluorobenzene
CAS Number:
Linear Formula:
 ClC6F5
Molecular Weight:
 202.51
 Beilstein/REAXYS Number:
 1819389
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More