• USA Home
  • Search Results

  • chlorophylla8934947961811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorophyll a

4 Products
Image of Chlorophyll a
CAS Number:
Empirical Formula (Hill Notation):
 C55H72MgN4O5
Molecular Weight:
 893.49
 Beilstein/REAXYS Number:
 3586237
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Special Grade
  Biological Source
  Assay
  Absorption
Show More