• USA Home
  • Search Results

  • chloroquinediphosphatesalt515865063511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chloroquine diphosphate salt

1 Products
Synonym:   N4-(7-Chloro-4-quinolinyl)-N1,N1-dimethyl-1,4-pentanediamine diphosphate salt, N4-(7-chloroquinolin-4-yl)-N1,N1-diethylpentane-1,4-diamine diphosphate
Image of Chloroquine diphosphate salt
CAS Number:
Empirical Formula (Hill Notation):
 C18H26ClN3 · 2H3PO4
Molecular Weight:
 515.86
 Beilstein/REAXYS Number:
 4223142
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Attribute Used:1/3