• USA Home
  • Search Results

  • chlorotrimethylsilane108647577411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorotrimethylsilane

7 Products
Synonym:   TMCS, TMSCl, Trimethylchlorosilane, Trimethylsilyl chloride
Image of Chlorotrimethylsilane
CAS Number:
Linear Formula:
 (CH3)3SiCl
Molecular Weight:
 108.64
 Beilstein/REAXYS Number:
 1209232
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Special Grade
  Vapor pressure
  Transition temperature
  Purification method
  Melting Point
  Explosion limit
  Density
  Autoignition temp
  Potency
Show More