• USA Home
  • Search Results

  • chlorotrimethylsilanesolution108647577411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorotrimethylsilane solution

1 Products
Synonym:   Trimethylchlorosilane
Image of Chlorotrimethylsilane solution
CAS Number:
Linear Formula:
 (CH3)3SiCl
Molecular Weight:
 108.64
 Beilstein/REAXYS Number:
 1209232
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Density
Show More