• USA Home
  • Search Results

  • chlortetracyclinehydrochloride515346472211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlortetracycline hydrochloride

7 Products
Synonym:   7-Chlorotetracycline hydrochloride
Image of Chlortetracycline hydrochloride
CAS Number:
Empirical Formula (Hill Notation):
 C22H23ClN2O8 · HCl
Molecular Weight:
 515.34
 Beilstein/REAXYS Number:
 3858364
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
  Solubility
  Product Line
  Assay
  Mode of Action
  Anal. Standard Form
  Special Grade
  pK-Values
  Physical Form
Show More