• USA Home
  • Search Results

  • chlorzoxazone169579525011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chlorzoxazone

3 Products
Synonym:   5-Chloro-2(3H)-benzoxazolone, 5-Chloro-2-hydroxybenzoxazole
Image of Chlorzoxazone
CAS Number:
Empirical Formula (Hill Notation):
 C7H4ClNO2
Molecular Weight:
 169.57
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
Show More