• USA Home
  • Search Results

  • cholecalciferol384646797011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cholecalciferol

5 Products
Synonym:   (+)-Vitamin D3, 7-Dehydrocholesterol activated, Activated 7-dehydrocholesterol, Calciol
Image of Cholecalciferol
CAS Number:
Empirical Formula (Hill Notation):
 C27H44O
Molecular Weight:
 384.64
 Beilstein/REAXYS Number:
 2339331
 EC Number:
 MDL number:
 NACRES:
 NA.26
 PubChem Substance ID:
 eCl@ss:
 34058003
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Agency/Method
  Anal. Standard Form
  Manufacturer designation
  Special Grade
  Color
  Biological Source
  Assay
Show More