• USA Home
  • Search Results

  • cholesta35diene3686474790011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cholesta-3,5-diene

1 Products
Image of Cholesta-3,5-diene
CAS Number:
Empirical Formula (Hill Notation):
 C27H44
Molecular Weight:
 368.64
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More