• USA Home
  • Search Results

  • cholesterol386655788511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cholesterol

8 Products
Synonym:   3β-Hydroxy-5-cholestene, 5-Cholesten-3β-ol
Image of Cholesterol
CAS Number:
Empirical Formula (Hill Notation):
 C27H46O
Molecular Weight:
 386.65
 Beilstein/REAXYS Number:
 1915888
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
 eCl@ss:
 39023139
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Package quantity
  Melting Point
  Geographic origin
  Agency/Method
  Anal. Standard Form
  Application(s)
  Manufacturer designation
  Special Grade
  Ice Shipping
  Assay
  Biological Source
  Physical Form
Show More