• USA Home
  • Search Results

  • cholesteryllinoleate6490860433111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cholesteryl linoleate

1 Products
Synonym:   3β-Hydroxy-5-cholestene 3-linoleate, 5-Cholesten-3β-ol 3-linoleate, Cholesteryl 9,12-octadecadienoate, Cholesteryl octadecadienoate
Image of Cholesteryl linoleate
CAS Number:
Empirical Formula (Hill Notation):
 C45H76O2
Molecular Weight:
 649.08
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More