• USA Home
  • Search Results

  • cholesterylpalmitate6250660134311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cholesteryl palmitate

2 Products
Synonym:   5-Cholestene 3-palmitate
Image of Cholesteryl palmitate
CAS Number:
Empirical Formula (Hill Notation):
 C43H76O2
Molecular Weight:
 625.06
 Beilstein/REAXYS Number:
 2342867
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Transition temperature
  Functional group
  Physical Form
  Ice Shipping
  Assay
Show More