• USA Home
  • Search Results

  • cholesterylstearate653123560269811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cholesteryl stearate

1 Products
Synonym:   5-Cholesten-3β-yl octadecanoate
Image of Cholesteryl stearate
CAS Number:
Empirical Formula (Hill Notation):
 C45H80O2
Molecular Weight:
 653.12
 Beilstein/REAXYS Number:
 2068492
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Transition temperature
Show More

Product #
Description
C79409 Cholesteryl stearate, 96%,