• USA Home
  • Search Results

  • cholicacid408578125411

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cholic acid

4 Products
Synonym:   3α,7α,12α-Trihydroxy-5β-cholanic acid, Cholanic acid
Image of Cholic acid
CAS Number:
Empirical Formula (Hill Notation):
 C24H40O5
Molecular Weight:
 408.57
 Beilstein/REAXYS Number:
 2822009
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Anal. Standard Form
  Special Grade
Show More