• USA Home
  • Search Results

  • cholinechloride139626748111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Choline chloride

9 Products
Synonym:   (2-Hydroxyethyl)trimethylammonium chloride
Image of Choline chloride
CAS Number:
Linear Formula:
 (CH3)3N(Cl)CH2CH2OH
Molecular Weight:
 139.62
 Beilstein/REAXYS Number:
 3563126
 EC Number:
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
  Anal. Standard Form
  Suitability
  UV-Absorption Values
  Molecular weight
  Solubility
  Agency/Method
  pH Value
  Product Line
  Assay
  Special Grade
  Biological Source
  Application(s)
  Color
Show More