• USA Home
  • Search Results

  • cholinehydroxidesolution1211812341111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Choline hydroxide solution

3 Products
Image of Choline hydroxide solution
CAS Number:
Linear Formula:
 HOCH2CH2N(CH3)3OH
Molecular Weight:
 121.18
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Concentration
  Refractive Index
  Density
Show More