• USA Home
  • Search Results

  • chondroitinsulfateasodiumsaltfrombovinetrachea123453945518011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chondroitin sulfate A sodium salt from bovine trachea

1 Products
Synonym:   Alternating Copoly β-glucuronic acid-(1→3)-N-acetyl-β-galactosamine-4-sulfate-(1→4)
Image of Chondroitin sulfate A sodium salt from bovine trachea
CAS Number:
 MDL number:
 NACRES:
 NA.75
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Biological Source
  Package quantity
Show More