• USA Home
  • Search Results

  • chromiumiiichloridehexahydrate266451006012511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chromium(III) chloride hexahydrate

3 Products
Synonym:   Chromic chloride hexahydrate, Chromium trichloride hexahydrate, Hexaaquachromium(III) chloride
Image of Chromium(III) chloride hexahydrate
CAS Number:
Linear Formula:
 CrCl3 · 6H2O
Molecular Weight:
 266.45
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Purity Grade
  Reaction Suitability
Show More