• USA Home
  • Search Results

  • chromiumvioxide9999133382011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chromium(VI) oxide

6 Products
Synonym:   Chromic anhydride
Image of Chromium(VI) oxide
CAS Number:
Empirical Formula (Hill Notation):
 CrO3
Molecular Weight:
 99.99
 EC Number:
 MDL number:
 NACRES:
 NA.55
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Assay
  Product Line
  Melting Point
  Density
  Bulk Density
  Restriction
  Special Grade
  Physical Form
  pH Value
Show More