• USA Home
  • Search Results

  • chromomycina3fromstreptomycesgriseus118325705924711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chromomycin A3 from Streptomyces griseus

1 Products
Image of Chromomycin A<SUB>3</SUB> from <I>Streptomyces griseus</I>
CAS Number:
Empirical Formula (Hill Notation):
 C57H82O26
Molecular Weight:
 1183.25
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Activity Spectrum
Show More