• USA Home
  • Search Results

  • chromotrope2r46837419707311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chromotrope 2R

1 Products
Synonym:   2-(Phenylazo)chromotropic acid disodium salt, Acid Red 29
Image of Chromotrope 2R
CAS Number:
Empirical Formula (Hill Notation):
 C16H10N2Na2O8S2
Molecular Weight:
 468.37
 Colour Index Number:
 16570
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
Show More