• USA Home
  • Search Results

  • chromotropefb50243356769911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chromotrope FB

1 Products
Synonym:   Acid Red 14, Disodium 4-hydroxy-3-[(4-sulfo-1-naphthalenyl)azo]-1-naphthalenesulfonate, Mordant Blue 79
Image of Chromotrope FB
CAS Number:
Empirical Formula (Hill Notation):
 C20H12N2Na2O7S2
Molecular Weight:
 502.43
 Colour Index Number:
 14720
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  RAMAN FTIR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Absorption
Show More