• USA Home
  • Search Results

  • chrysin2542448040011

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Chrysin

3 Products
Synonym:   5,7-Dihydroxyflavone
Image of Chrysin
CAS Number:
Empirical Formula (Hill Notation):
 C15H10O4
Molecular Weight:
 254.24
 Beilstein/REAXYS Number:
 233276
 EC Number:
 MDL number:
 NACRES:
 NA.47
 PubChem Substance ID:
Popular Documents:  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Application(s)
  Physical Form
  Special Grade
  Assay
Show More