• USA Home
  • Search Results

  • ciclesonide5406912654447611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Ciclesonide

2 Products
Image of Ciclesonide
CAS Number:
Empirical Formula (Hill Notation):
 C32H44O7
Molecular Weight:
 540.69
 MDL number:
 NACRES
 NA.24
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Solubility
  Physical Form
  Color
Show More