• USA Home
  • Search Results

  • cimetidine252345148161911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cimetidine

5 Products
Synonym:   SKF-92334
Image of Cimetidine
CAS Number:
Empirical Formula (Hill Notation):
 C10H16N6S
Molecular Weight:
 252.34
 EC Number:
 MDL number:
 NACRES:
 NA.77
 PubChem Substance ID:
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Anal. Standard Form
  Physical Form
  Application(s)
  Manufacturer designation
Show More