• USA Home
  • Search Results

  • cimetidineforsystemsuitability252345148161911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cimetidine for system suitability

1 Products
Synonym:   Cimetidine, SKF-92334
Image of Cimetidine for system suitability
CAS Number:
Empirical Formula (Hill Notation):
 C10H16N6S
Molecular Weight:
 252.34
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More