• USA Home
  • Search Results

  • cinchonine2943911810511

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cinchonine

1 Products
Synonym:   NSC 6176
Image of Cinchonine
CAS Number:
Empirical Formula (Hill Notation):
 C19H22N2O
Molecular Weight:
 294.39
 Beilstein/REAXYS Number:
 89689
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More