• USA Home
  • Search Results

  • cinchoninemonohydrochloridehydrate33085anhydrousbasis20698688111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cinchonine monohydrochloride hydrate

1 Products
Synonym:   Cinchonan-9(S)-ol monohydrochloride
Image of Cinchonine monohydrochloride hydrate
CAS Number:
Empirical Formula (Hill Notation):
 C19H22N2O · HCl · xH2O
Molecular Weight:
 330.85 (anhydrous basis)
 Beilstein/REAXYS Number:
 5784953
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More