• USA Home
  • Search Results

  • cineole1542547082611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cineole

1 Products
Synonym:   1,3,3-Trimethyl-2-oxabicyclo[2.2.2]octane, 1,8-Cineole, 1,8-Epoxy-p-menthane, Eucalyptol
Image of Cineole
CAS Number:
Empirical Formula (Hill Notation):
 C10H18O
Molecular Weight:
 154.25
 Beilstein/REAXYS Number:
 105109
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More