• USA Home
  • Search Results

  • cinnamaldehyde1321610455211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cinnamaldehyde

2 Products
Synonym:   3-Phenylprop-2-enal, Cinnamal, Cinnamic aldehyde
Image of Cinnamaldehyde
CAS Number:
Linear Formula:
 C6H5CH=CHCHO
Molecular Weight:
 132.16
 Council of Europe no.:
 102c
 EC Number:
 FEMA Number:
 2286
 Flavis number:
 5.014
 MDL number:
 NACRES:
 NA.21
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Agency/Method
  Special Grade
  Assay
Show More