• USA Home
  • Search Results

  • cinnamicacid1481614010311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cinnamic acid

3 Products
Synonym:   (2E)-3-Phenyl-2-propenoic acid, trans-3-Phenylacrylic acid, trans-Cinnamic acid
Image of Cinnamic acid
CAS Number:
Empirical Formula (Hill Notation):
 C9H8O2
Molecular Weight:
 148.16
 Beilstein/REAXYS Number:
 1905952
 MDL number:
 NACRES:
 NA.24
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Potency
Show More