• USA Home
  • Search Results

  • cinnamonbarkoil12345801591611

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cinnamon bark oil

1 Products
Synonym:   Cassia oil, Cinnamomum
CAS Number:
 FEMA Number:
 2291
 MDL number:
 NACRES:
 NA.21
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Refractive Index
  Document Links
  Allergen
  Organoleptic property
  Density
  Biological Source
  Agency/Method
Show More