• USA Home
  • Search Results

  • cinnamylacetate1762110354811

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cinnamyl acetate

4 Products
Image of Cinnamyl acetate
CAS Number:
Linear Formula:
 CH3CO2CH2CH=CHC6H5
Molecular Weight:
 176.21
 EC Number:
 MDL number:
 NACRES:
 NA.22
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Special Grade
  Document Links
  Allergen
  Organoleptic property
  Application(s)
  Solubility
  Density
  Assay
  Agency/Method
Show More