• USA Home
  • Search Results

  • cinnarizine3685129857711

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cinnarizine

2 Products
Synonym:   1-trans-Cinnamyl-4-diphenylmethylpiperazine
Image of Cinnarizine
CAS Number:
Empirical Formula (Hill Notation):
 C26H28N2
Molecular Weight:
 368.51
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Physical Form
Show More