• USA Home
  • Search Results

  • cinoxacin262222865780911

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

Cinoxacin

1 Products
Synonym:   1-Ethyl-1,4-dihydro-4-oxo-[1,3]dioxolo[4,5-g]cinnoline-3-carboxylic acid
Image of Cinoxacin
CAS Number:
Empirical Formula (Hill Notation):
 C12H10N2O5
Molecular Weight:
 262.22
 EC Number:
 MDL number:
 NACRES:
 NA.85
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.

Available Attributes

Use the product Attributes below to configure the comparison table. (Select up to %1% total)
  Brand
  Mode of Action
Show More

Product #
Description
C8645 Cinoxacin,