• USA Home
  • Search Results

  • cis12dichloroethylene969415659211

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-1,2-Dichloroethylene

1 Products
Synonym:   cis-1,2-DCE, cis-Acetylene dichloride
Image of <I>cis</I>-1,2-Dichloroethylene
CAS Number:
Linear Formula:
 ClCH=CHCl
Molecular Weight:
 96.94
 Beilstein/REAXYS Number:
 1071208
 EC Number:
 MDL number:
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)  |  FTNMR (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More