• USA Home
  • Search Results

  • cis1316docosadienoicacidmethylester350586101247311

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-13,16-Docosadienoic acid methyl ester

1 Products
Image of <I>cis</I>-13,16-Docosadienoic acid methyl ester
CAS Number:
Empirical Formula (Hill Notation):
 C23H42O2
Molecular Weight:
 350.58
 MDL number:
 NACRES:
 NA.25
 PubChem Substance ID:
Popular Documents:  Specification Sheet (PDF)
  
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More