• USA Home
  • Search Results

  • cis2butene561159018111

EMAIL THIS PAGE TO A FRIEND

Product Comparison Guide

cis-2-Butene

1 Products
Image of <I>cis</I>-2-Butene
CAS Number:
Linear Formula:
 CH3CH=CHCH3
Molecular Weight:
 56.11
 Beilstein/REAXYS Number:
 1361341
 EC Number:
 MDL number:
 PubChem Substance ID
You have selected the maximum number of product attributes (%1%) to compare. Please remove at least one to add a new selection.
Show More

Product #
Description
400890 cis-2-Butene, ≥99%,